missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Histone H1.5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA525155
This item is not returnable.
View return policy
Description
This antibody is predicted to react with rabbit based on sequence homology.
Histones are basic nuclear proteins responsible for nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H1 family.
Specifications
| Histone H1.5 | |
| Polyclonal | |
| Unconjugated | |
| H1-5 | |
| H1; H1 histone family; H1 histone family, member 5; H1.5; H1B; H1F5; H1s-3; HIST1H1B; histone 1, H1b; histone cluster 1, H1b; histone H1.5; Histone H1a; histone H1b; Histone H1s-3; MGC126630; MGC126632 | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 3009 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P16401 | |
| H1-5, HIST1H1B | |
| KLH conjugated synthetic peptide between 33-60 amino acids from the N-terminal region of human HIST1H1B. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction