Learn More
MedChemExpress Hinokitiol
Specifications
Specifications
| Content And Storage | Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month |
| Molecular Weight (g/mol) | 164.2 g/mol |
| Shipping Condition | Room temperature |
| For Use With (Application) | Cancer-programmed cell death |
| CAS | 499-44-5 |
| Product Type | Natural Products |
| Purity | 98.0% |
| Solubility | DMSO : 100 mg/mL (609.01 mM∣Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) |
| Synonym | β-Thujaplicin |
| Target | Keap1-Nrf2∣DNA Methyltransferase∣Virus Protease |
| Show More |