Learn More
Porcelaines Avignon™ High Form Porcelain Crucibles
Product Code. 11728483
Click to view available options
Capacity (Metric):
40 mL
Unit Size:
Pack of 6
Product Code. 11728483
Brand: Porcelaines Avignon™ 11206
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
High form, glazed porcelain crucible with a high resistance to thermal and physical shock. Porcelaines Avignon™ High Form Porcelain Crucibles are chemically neutral with an enamel that is unalterable to most concentrated acids.
  • Specialized porcelain with a very low expansion coefficient.
  • High resistance to thermal and physical shocks of all types.
  • Glazed porcelain - can withstand temperatures up to 1200°C
  • Liquid and gas tight.
  • Chemically neutral.
  • Enamel is unalterable to concentrated acids (with the exception of hydrofluric acid), even in the boiling state and for an almost unlimited duration.
Specifications
Material Porcelain
Finish Glazed
Max. Temperature (Metric) 1200°C
Capacity (Metric) 40 mL
Diameter (Metric) Outer 45mm
Height (Metric) 51mm
Product Type High Form Porcelain Crucibles