Learn More
Agilent BioTek High Flow Complete Dispense/Waste System
Shop All Agilent BioTek Products
Click to view available options
Capacity (Metric):
4 L
10 L
20 L
Unit Size:
Each
Description
- Includes four dispense bottles, two waste bottles and high flow vacuum pump
Specifications
Specifications
| Includes | 4 x 10 L Dispense Bottles, 2 x 10 L Waste Bottles, High Flow Vacuum Pump |
| Capacity (Metric) | 10 L |
| Type | Waste System |
| For Use With (Equipment) | 406 FX Washer Dispenser |