missing translation for 'onlineSavingsMsg'
Learn More

HIF-2 alpha/EPAS1 Antibody (2444A) - BSA Free, Novus Biologicals™

Product Code. 18268358 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.10mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18268358 0.1 mg 0.10mg
18360760 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18268358 Supplier Novus Biologicals Supplier No. NBP276454

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

HIF-2 alpha/EPAS1 Monoclonal specifically detects HIF-2 alpha/EPAS1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIF-2 alpha/EPAS1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2444A
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunohistochemistry-Paraffin
Formulation PBS with 0.02% Sodium Azide
Gene Alias anti hif 2 alpha, anti-HIF-2 alpha, Basic-helix-loop-helix-PAS protein MOP2, BHLHE73, Class E basic helix-loop-helix protein 73, ECYT4, endothelial PAS domain protein 1, endothelial PAS domain-containing protein 1, ep190b, EPAS1, EPAS-1, HIF-1-alpha-like
Gene Symbols EPAS1
Host Species Rabbit
Immunogen This recombinant HIF-2 alpha/EPAS1 Antibody (2444A) is made to a partial recombinant human HIF-2 protein (amino acids 543-870) [UniProt Q99814]
Molecular Weight of Antigen 96.5 kDa
Purification Method Protein A or G purified
Quantity 0.1 mg
Research Discipline Angiogenesis, Apoptosis, Cancer, Cancer Stem Cells, Chromatin Research, Embryonic Stem Cell Markers, HIF Target Genes, Hypoxia, Lipid and Metabolism, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 2034.0
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.