missing translation for 'onlineSavingsMsg'
Learn More

HG6-64-1, MedChemExpress

Product Code. 16485068
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16485068

Brand: Medchem Express HY12291

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress HG6-64-1 is a potent and selective B-Raf inhibitor extracted from patent WO 2011090738 A2, example 9 (XI-1); has a IC50 of 0.09 μM on B-raf V600E transformed Ba/F3 cells.

HG6-64-1 is a potent and selective B-Raf inhibitor extracted from patent WO 2011090738 A2, example 9 (XI-1)

Specifications

Chemical Name or Material HG6-64-1
Physical Form Solid
CAS 1315329-43-1
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C32H34F3N5O2
For Use With (Application) Cancer-Kinase/protease
Synonym HMSL 10017-101-1
Solubility Information DMSO : ≥ 100 mg/mL (173.12 mM)
SMILES O=C(NC1=CC=C(CN2CCN(CC)CC2)C(C(F)(F)F)=C1)C3=CC=C(C)C(/C=C/C4=CN=C(NC=C5)C5=C4OC)=C3
Molecular Weight (g/mol) 577.64
Formula Weight 577.64
Percent Purity 96.37%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.