Learn More
HG-14-10-04, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
HG-14-10-04 (example 10) is a potent and specific ALK inhibitor with an IC50 of 20 nM.
Specifications
Specifications
| Chemical Name or Material | HG-14-10-04 |
| CAS | 1356962-34-9 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C29H34ClN7O |
| Quantity | 100 mg |
| Solubility Information | DMSO : 20 mg/mL (37.59 mM; Need ultrasonic) |
| SMILES | ClC1=CN=C(NC2=CC=C(N3CCC(N4CCN(C)CC4)CC3)C=C2OC)N=C1C5=CNC6=CC=CC=C65 |
| Molecular Weight (g/mol) | 532.08 |
| Formula Weight | 532.08 |
| Show More |