Learn More
Hexoprenaline Sulfate, TRC
Specifications
Specifications
| Chemical Name or Material | Hexoprenaline Sulfate |
| CAS | 32266-10-7 |
| Molecular Formula | C22H32N2O6 . H2SO4 |
| Quantity | 10 mg |
| Synonym | 4,4'-[1,6-hexanediylbis[imino(1-hydroxy-2,1-ethanediyl)]]bis-1,2-Benzenediol sulfate (1:1) (salt), Asmadol, Bronalin, Delaprem, Etoscol, Gynebral, Gynipral, Leanol, NSC 292266, ST 1512/SO4 |
| InChI Formula | InChI=1S/C22H32N2O6.H2O4S/c25-17-7-5-15(11-19(17)27)21(29)13-23-9-3-1-2-4-10-24-14-22(30)16-6-8-18(26)20(28)12-16;1-5(2,3)4/h5-8,11-12,21-30H,1-4,9-10,13-14H2;(H2,1,2,3,4) |
| SMILES | OC(CNCCCCCCNCC(O)c1ccc(O)c(O)c1)c2ccc(O)c(O)c2.OS(=O)(=O)O |
| IUPAC Name | 4-[2-[6-[[2-(3,4-dihydroxyphenyl)-2-hydroxyethyl]amino]hexylamino]-1-hydroxyethyl]benzene-1,2-diol;sulfuric acid |
| Molecular Weight (g/mol) | 518.6 |
| Formula Weight | 518.1934 |