Learn More
Hexoprenaline Sulfate, TRC
Product Code. 30393598
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393598 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30393598 Supplier Toronto Research Chemicals Supplier No. TRCH29546350MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Hexoprenaline Sulfate
CAS 32266-10-7
Molecular Formula C22H32N2O6 . H2SO4
Quantity 50 mg
Synonym 4,4'-[1,6-hexanediylbis[imino(1-hydroxy-2,1-ethanediyl)]]bis-1,2-Benzenediol sulfate (1:1) (salt), Asmadol, Bronalin, Delaprem, Etoscol, Gynebral, Gynipral, Leanol, NSC 292266, ST 1512/SO4
InChI Formula InChI=1S/C22H32N2O6.H2O4S/c25-17-7-5-15(11-19(17)27)21(29)13-23-9-3-1-2-4-10-24-14-22(30)16-6-8-18(26)20(28)12-16;1-5(2,3)4/h5-8,11-12,21-30H,1-4,9-10,13-14H2;(H2,1,2,3,4)
SMILES OC(CNCCCCCCNCC(O)c1ccc(O)c(O)c1)c2ccc(O)c(O)c2.OS(=O)(=O)O
IUPAC Name 4-[2-[6-[[2-(3,4-dihydroxyphenyl)-2-hydroxyethyl]amino]hexylamino]-1-hydroxyethyl]benzene-1,2-diol;sulfuric acid
Molecular Weight (g/mol) 518.6
Formula Weight 518.1934