missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexokinase 1 Antibody [Allophycocyanin], Novus Biologicals Biologicals™
Shop All Bio Techne ProductsDescription
Hexokinase 1 Polyclonal antibody specifically detects Hexokinase 1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | Hexokinase 1 |
| Applications | ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | APC |
| Formulation | PBS |
| Gene Alias | Brain form hexokinase, EC 2.7.1, EC 2.7.1.1, glycolytic enzyme, hexokinase 1, Hexokinase type I, hexokinase-1, HK I, HK1-ta, HK1-tb, HK1-tc, HKI, HXK1 |
| Host Species | Rabbit |
| Immunogen | Hexokinase 1 antibody was raised against a 16 amino acid peptide near the carboxy terminus of human Hexokinase 1. The immunogen is located within amino acids 830 - 880 of Hexokinase 1. |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?