Learn More
Hexestrol Dimethyl Ether-d6, TRC
Product Code. 30404534
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30404534 1 mg 1mg
30399839 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30404534 Supplier Toronto Research Chemicals Supplier No. TRCH2953121MG
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Hexestrol Dimethyl Ether-d6
Molecular Formula C20 D6 H20 O2
Quantity 1 mg
Synonym 3,4-Bis(4-methoxyphenyl)hexane-d6, 3,4-Di(p-methoxyphenyl)hexane-d6, alpha,alpha'-Diethyl-4,4'-dimethoxybibenzyl-d6, 1,1'-(1,2-Diethyl-1,2-ethanediyl)bis[4-methoxybenzene-d6
InChI Formula InChI=1S/C20H26O2/c1-5-19(15-7-11-17(21-3)12-8-15)20(6-2)16-9-13-18(22-4)14-10-16/h7-14,19-20H,5-6H2,1-4H3/i1D3,2D3
SMILES [2H]C([2H])([2H])CC(C(CC([2H])([2H])[2H])c1ccc(OC)cc1)c2ccc(OC)cc2
IUPAC Name 1-[1,1,1,6,6,6-hexadeuterio-4-(4-methoxyphenyl)hexan-3-yl]-4-methoxybenzene
Molecular Weight (g/mol) 304.456
Formula Weight 304.231