missing translation for 'onlineSavingsMsg'
Learn More

Hexestrol, 98+%, Thermo Scientific Chemicals

Product Code. 15483579
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15483579 10 g 10g
15473579 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15483579 Supplier Thermo Scientific Alfa Aesar Supplier No. J60889.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Nonsteroidal synthetic estrogen

Nonsteroidal synthetic estrogen

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-16-2
Molecular Formula C18H22O2
Molecular Weight (g/mol) 270.37
MDL Number MFCD00068996
InChI Key PBBGSZCBWVPOOL-UHFFFAOYNA-N
Synonym hexestrol, meso-hexestrol, hexoestrol, hexanoestrol, hexestrofen, mesohexestrol, erythrohexestrol, estra-plex, cycloestrol, estrifar
PubChem CID 192197
SMILES CCC(C(CC)C1=CC=C(O)C=C1)C1=CC=C(O)C=C1

Specifications

Melting Point 186°C
Color White
Quantity 10 g
Beilstein 3209460
Merck Index 14,4701
IUPAC Name 4-[4-(4-hydroxyphenyl)hexan-3-yl]phenol
Formula Weight 270.37
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Hexestrol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.