missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HEXB Polyclonal Antibody
GREENER_CHOICE

Product Code. 15751012 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15751012 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15751012 Supplier Invitrogen™ Supplier No. PA563236

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: FGFYKWHHEP AEFQAKTQVQ QLLVSITLQS ECDAFPNISS DESYTLLVKE PVAVLKANRV W Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 69%.

Hexosaminidase B (HEXB), also designated beta-hexosaminidase B, is a Hexosaminidase B (HEXB), also designated b-hexosaminidase B, is a tetramer of two b-A and two b-B chains and is found in the lysosomes of cells. Sandhoff disease (SD), also known as GM2-gangliosidosis type II, is caused by mutations in the HEXB gene that affect the b subunit. These mutations disrupt the activity of HEXB and HEXA, which prevents the breakdown of GM2 ganglioside, a fatty material found in the brain, therby rendering both the HEXA and HEXB enzymes deficient. SD is a rare autosomal recessive disorder characterized by an accumulation of GM2 ganglioside, which causes progressive destruction of the central nervous system. Sandhoff disease is similar to Tay-Sachs disease, which is caused by mutations in the HEXA gene, although SD is more severe.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEXB
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Hexb
Gene Accession No. P07686
Gene Alias beta-hexosaminidase subunit beta; Beta-hexosaminidase subunit beta chain A; Beta-hexosaminidase subunit beta chain B; Beta-N-acetylhexosaminidase subunit beta; cervical cancer proto-oncogene 7 protein; ENC-1AS; epididymis luminal protein 248; epididymis secretory protein Li 111; HCC7; HCC-7; HEL-248; HEL-S-111; HEXB; hexosaminidase B; hexosaminidase B (beta polypeptide); hexosaminidase subunit B; hexosaminidase subunit beta; N-acetyl-beta-glucosaminidase subunit beta
Gene Symbols Hexb
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HEXB. Recombinant protein control fragment (Product #RP-100907).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3074
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.