Learn More
Hexanedioyl-d8 Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | Hexanedioyl Dichloride-D8 |
| CAS | 284487-62-3 |
| Molecular Formula | C6 D8 Cl2 O2 |
| Quantity | 5 mg |
| Synonym | Hexanedioyl-d8 dichloride (9CI), Hexanedioyl-d8 chloride, Hexanedioyl dichloride-D8, Adipoyl chloride-d8, Hexanedioic dichloride-d8, Adipic acid dichloride-d8 |
| InChI Formula | InChI=1S/C6H8Cl2O2/c7-5(9)3-1-2-4-6(8)10/h1-4H2/i1D2,2D2,3D2,4D2 |
| SMILES | [2H]C([2H])(C(=O)Cl)C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)Cl |
| IUPAC Name | 2,2,3,3,4,4,5,5-octadeuteriohexanedioyl dichloride |
| Molecular Weight (g/mol) | 191.0818 |
| Formula Weight | 190.0403 |