missing translation for 'onlineSavingsMsg'
Learn More

gamma-Hexalactone, 98%

Product Code. p-7077259 Shop All Thermo Scientific Products
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 15595086

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 695-06-7 | C6H10O2 | 114.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 695-06-7
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00005401
InChI Key JBFHTYHTHYHCDJ-UHFFFAOYNA-N
Synonym gamma-caprolactone, gamma-hexalactone, 4-hexanolide, gamma-hexanolactone, 6-caprolactone, tonkalide, toukalide, 2 3h-furanone, 5-ethyldihydro, 4-ethyl-4-butanolide, 5-ethyldihydrofuran-2 3h-one
PubChem CID 12756
ChEBI CHEBI:85235
IUPAC Name 5-ethyloxolan-2-one
SMILES CCC1CCC(=O)O1

Specifications

Melting Point -18.0°C
Density 1.0230g/mL
Boiling Point 220.0°C
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 25 g
Specific Gravity 1.023
Solubility Information Solubility in water: slightly soluble.
Formula Weight 114.14
Percent Purity 98%
Chemical Name or Material γ-Hexalactone
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.