Learn More
Heptanal-d14, TRC
Product Code. 30395370
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395370 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30395370 Supplier Toronto Research Chemicals Supplier No. TRCH28114225MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Heptanal-d14
CAS 1246819-97-5
Molecular Formula C7 D14 O
Quantity 25 mg
Synonym 1,2,2,3,3,4,4,5,5,6,6,7,7,7-Tetradecadeuterioheptan-1-one, Heptanal-d14, Heptanal-D14, Heptanal-1,2,2,3,3,4,4,5,5,6,6,7,7,7-d14, 1-Heptaldehyde-d14, Enanthal-d14, Enanthaldehyde-d14, Enanthic Aldehyde-d14, Enanthole-d14, Heptaldehyde-d14, Heptanaldehyde-d14, Heptyl Aldehyde-d14, Oenanthal-d14, Oenanthaldehyde-d14, Oenanthic Aldehyde-d14, Oenanthol-d14, n-Heptaldehyde-d14, n-Heptanal-d14, n-Heptylaldehyde-d14
InChI Formula InChI=1S/C7H14O/c1-2-3-4-5-6-7-8/h7H,2-6H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D
SMILES [2H]C(=O)C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]
IUPAC Name 1,2,2,3,3,4,4,5,5,6,6,7,7,7-tetradecadeuterioheptan-1-one
Molecular Weight (g/mol) 128.2717
Formula Weight 128.1923