missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Heptamethine cyanine dye-1

Product Code. 16441198
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16441198 5 mg 5mg
16478768 10 mg 10mg
16454687 50 mg 50mg
16428916 100 mg 100mg
16403826 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16441198 Supplier MedChemExpress Supplier No. HYD0921
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Heptamethine cyanine dye-1 is a near-infrared cyanine dye for fluorescence imaging in biological systems.

Specifications

Quantity 5 mg
Product Type Dye Reagents
Molecular Weight (g/mol) 739.17 g/mol
Solubility DMSO : 50 mg/mL (67.64 mM; Need ultrasonic)
Content And Storage 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Shipping Condition Room temperature
Target HBV
CAS 162411-29-2
Synonym ADS 815EI
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.