Learn More
Heptadecafluorooctanesulfonic Acid Potassium, TRC
Specifications
Specifications
| Chemical Name or Material | Perfluorooctane sulfonic acid potassium salt (PFOS) |
| CAS | 2795-39-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C8 F17 O3 S . K |
| Quantity | 10 mg |
| Synonym | Perfluorooctane Sulfonic Acid Potassium Salt (PFOS), Potassium perfluorooctanesulfonate, 1-Octanesulfonic acid, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-, potassium salt (8CI, 9CI), 1-Octanesulfonic acid, heptadecafluoro-, potassium salt (6CI), EF 102, Eftop EF 102, F 110, F 116, FC 80, FC 95, FC 95 surfactant, Fluorad 95, Fluorad FC 95, Fluortensid FT 800, Heptadecafluorooctanesulfonic acid potassium salt, LE 003, Megafac F 110, Megafac F 116, Perfluorooctanesulfonic acid potassium salt, Potassium heptadecafluorooctan-1-sulfonate, Potassium heptadecafluorooctane-1-sulfonate, Potassium heptadecafluorooctanesulfonate, Potassium perfluorooctanesulfonate, Potassium perfluorooctylsulfonate, YF 95 |
| InChI Formula | InChI=1S/C8HF17O3S.K/c9-1(10,3(13,14)5(17,18)7(21,22)23)2(11,12)4(15,16)6(19,20)8(24,25)29(26,27)28;/h(H,26,27,28);/q;+1/p-1 |
| SMILES | [K+].[O-]S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
| IUPAC Name | potassium;1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane-1-sulfonate |
| Molecular Weight (g/mol) | 538.22 |
| Show More |