missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Hepatocyte Thaw Medium

Product Code. 15233953
Change view
Click to view available options
Quantity:
45 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15233953 45 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15233953 Supplier Gibco™ Supplier No. CM7500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hepatocyte Thaw Medium

Gibco Hepatocyte Thaw Medium is a ready-to-use medium specifically designed for use with thawed cryopreserved hepatocytes to enhance their viability and overall quality.

Features of Hepatocyte Thaw Medium:

  • Recover cryopreserved hepatocytes with a single centrifugation
  • Designed for use with Gibco human and animal cryopreserved hepatocytes for optimal results
  • Provides high quality hepatocytes suitable for many in vitro assays, including induction, metabolic stability, and transporter assessments

Hepatocyte Thaw Medium is easy to use. Simply warm the bottle of medium to 37°C prior to thawing the cryopreserved hepatocytes. Once the medium is warm, transfer it into a 50-mL centrifuge tube. Once the hepatocytes are thawed, pour the cells into the pre-warmed Hepatocyte Thaw Medium and centrifuge. Remove the supernatant and proceed with the next steps of your experiment. There is no need to centrifuge the hepatocytes a second time.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2–8°C) and protect from light.
Cell Type Hepatocyte Cells
Classification Animal Origin
Form Liquid
Product Type Cell Culture Supplement
Serum Level Standard Serum
With Additives Phenol Red
Format 50 mL Bottle
Quantity 45 mL
Source Animal origin, Human
Species Cattle/Bovine
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.