missing translation for 'onlineSavingsMsg'
Learn More

Helios Antibody (736440) [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30159673 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30159673 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30159673 Supplier Novus Biologicals Supplier No. FAB73092JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Helios Monoclonal antibody specifically detects Helios in Human samples. It is validated for Flow Cytometry (Intracellular Staining),CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Helios
Applications Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 736440
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias HELIOS, IKAROS family zinc finger 2 (Helios), Ikaros family zinc finger protein 2, MGC34330, subfamily 1A, 2 (Helios), zinc finger DNA binding protein Helios, zinc finger protein Helios, ZNF1A2, ZNFN1A2
Host Species Mouse
Immunogen E. coli-derived recombinant human Helios, Met1-Gln97, Accession # Q9UKS7
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 22807
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.