missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Helicobacter Pylori IgM ELISA Kit (Colorimetric)
Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Specifications
Specifications
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Serum |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Storage Requirements | Store at 4°C. |
| Synonym | H. pylori |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction