Learn More
Heidolph™ Instruments Hei-VAP Ultimate Control ML G5, EU-plug
The Hei-VAP Ultimate Control ML G5 adds numerous smart features to the Hei-VAP Ultimate.
Brand:  Heidolph™ Instruments 576-02500-00
Additional Details : Weight : 20.52000kg
Product Code. 17383370
  • 6815.00 € / Each
Estimated Shipment: 12-07-2024
to see stock.
Add to basket
Description
Description
Adds numerous smart features to the Hei-VAP Ultimate, as well as an overview of all parameters and the current process.
  • 7 in. touch display for controlling the entire system including the vacuum and the cooling temperature
  • Motor lift model with G5 dry ice condenser
  • For precise monitoring, the pictogram on the display also symbolizes active functions
  • Separate knobs for quick access: the right one for direct vacuum control, the left one for the rotation
  • Setting of values either via the touch display or the separate quick access knobs
  • Direct access to two favorites on the start screen
  • Expandable solvent library. You can also transfer your own favorites to other devices using a USB stick
  • Define and save own evaporation processes (ramps)
  • Automatically evaporate with the Dynamic AUTOaccurate Program (DAA). Convenient and customizable: from foam avoidance to highly efficient evaporation
  • USB and Micro SD plus LAN / RS 232 interfaces (Lab 4.0 ready)
Specifications
Specifications
5 L
Motor Lift Rotary Evaporator
EU-plug
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers