Description
Adds numerous smart features to the Hei-VAP Ultimate, as well as an overview of all parameters and the current process.- 7 in. touch display for controlling the entire system including the vacuum and the cooling temperature
- Hand lift model with transparent plastic coated G6 vertical glassware for reflux distillation
- For precise monitoring, the pictogram on the display also symbolizes active functions
- Separate knobs for quick access: the right one for direct vacuum control, the left one for the rotation
- Setting of values either via the touch display or the separate quick access knobs
- Direct access to two favorites on the start screen
- Expandable solvent library. You can also transfer your own favorites to other devices using a USB stick
- Define and save own evaporation processes (ramps)
- Automatically evaporate with the Dynamic AUTOaccurate Program (DAA). Convenient and customizable: from foam avoidance to highly efficient evaporation
- USB and Micro SD plus LAN / RS 232 interfaces (Lab 4.0 ready)
Specifications
5 L | |
Hand Lift Rotary Evaporator | |
EU-plug |