missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Heat Stable FGF-10 Recombinant Protein

Product Code. 17825410
Click to view available options
Quantity:
1 mg
100 μg
5 μg
50 μg
500 μg
Unit Size:
100µg
1mg
500µg
50µg
5µg
This item is not returnable. View return policy

Product Code. 17825410

Brand: Gibco™ PHG0371

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

FGF10 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein exhibits mitogenic activity for keratinizing epidermal cells, but essentially no activity for fibroblasts, which is similar to the biological activity of FGF7. Studies of the mouse homolog of suggested that this gene is required for embryonic epidermal morphogenesis including brain development, lung morphogenesis, and initiation of lim bud formation. This protein is also implicated to be a primary factor in the process of wound healing.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O15520
For Use With (Application) Functional Assay
Formulation protein with 20 mM Na2PO4, 750 mM NaCl and no preservative; pH 7.5
Gene ID (Entrez) 2255
Name Heat Stable FGF-10
Purification Method SDS-Page
Quantity 5 μg
Endotoxin Concentration 0.25 EU/μg
Gene Alias AEY17; BB213776; FGF; Fgf10; FGF-10; Fibroblast growth factor; Fibroblast growth factor 10; Gsfaey17; Keratinocyte growth factor 2; produced by fibroblasts of urinary bladder lamina propria
Common Name FGF10
Biological Activity ED50 = 1.0 ng/mL; measured in a cell proliferation assay using 4 MBr-5 cells
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Sequence Thermostable variant of human FGF-10 (190 amino acids) with proprietary amino acid substitutions
Buffer 750 mM NaCl, 20 mM Na2PO4
Content And Storage -20°Cor to +80°C if preferred
Classification Recombinant
Activity ED50 ≤1.0 ng/mL; measured in a cell proliferation assay using 4 MBr-5 cells
Expression System E. coli
Form Lyophilized
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.