Learn More
Brady™ Heat-shrink Polyolefin Wiremarker Sleeves
Specifications
Specifications
| Certifications/Compliance | RoHS compliant B-342 to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC, UL Recognized to UL224, SAE-AMS-DTL-23053/5 C (Class 1), SAE-AS-81531 |
| Description | BMP71 PermaSleeve Wire Marker Sleeves |
| Color | Yellow |
| Finish | Matte |
| Format | Roll |
| Material | Polyolefin |
| Printable Sides | Single-side Printable |
| Print Technology | Dot Matrix, Thermal Transfer |
| Product Type | Wire Marker Sleeve |
| For Use With (Application) | For wire and cable marking |
| Show More |