Learn More
Grant Instruments™ Heat Exchange Cooling Coil
Product Code. 11486487
Change view
Click to view available options
For Use With (Equipment):
OLS26
T100, TC120, TX150, TXF200
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11486487 T100, TC120, TX150, TXF200 Each
15345872 OLS26 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11486487 Supplier Grant Instruments™ Supplier No. CW5
Only null left
Add to basket
Add to basket
Can be attached to a cold water supply or refrigerated circulator. Grant Instruments™ Heat Exchange Coil maintains bath down to 2°C above the temperature of the coolant.
  • Stainless steel heat exchange coil that can be attached to cold water supply or a refrigerated circulator
  • Can be used to maintain a water bath temperature that is 2°C above the temperature of the coolant used
  • Compatible with OLS 26 Aqua Pro
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) T100, TC120, TX150, TXF200
Temperature Range 2°C
Type Heat Exchange Coil