Learn More
Kautex™ HDPE Wrench
Click to view available options
Thread Size (Metric):
00 mm
Unit Size:
Each
Description
- Wrench suitable for all sizes of wide neck closures from 100-4.000 mL
Specifications
Specifications
| For Use With (Application) | Closures |
| Material | HDPE |
| Thread Size (Metric) | 00 mm |
| Certified | Yes |
| Neck Finish | 00 mm |
| Certifications/Compliance | DIN 28 and DIN 80 |
| Product Type | Wrench |