Learn More
Masterflex™ HDPE T Connector
missing translation for 'orderingAttributeHoverText'
For Use With:
4.76mm
9.52mm
15.8mm
19.05mm
25.4mm
missing translation for 'unitSize'
Pack of 10
Spezifikation
Spezifikation
| Diameter (Metric) Inner | 9.52 mm |
| Material | HDPE |
| For Use With | 9.52mm |
| Min. Operating Temperature (Metric) | -53°C |
| Packaging Quantity | 10 Pack |
| Shape | T |
| Max. Operating Temperature (Metric) | 65°C |
| Product Type | T-Connector |