missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDLBP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | HDLBP |
|---|---|
| Concentration | 0.55 mg/mL |
| Applications | Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16834564
|
Proteintech
15406-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16824564
|
Proteintech
15406-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The function of this protein remains unknown.Specifications
| HDLBP | |
| Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| Q00341, Q8VDJ3, Q9Z1A6 | |
| 110611, 3069, 64474 | |
| HDLBP Fusion Protein Ag7666 | |
| Primary | |
| -20°C | |
| HDLBP |
| 0.55 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| HBP, HDL binding protein, HDLBP, PRO2900, VGL, Vigilin | |
| Hdlbp | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title