Learn More
HDHD1 Monoclonal Antibody (OTI7A2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502555
Description
This gene encodes a member of the haloacid dehalogenase-like (HAD) hydrolase superfamily. The encoded protein has no known biological function. This gene has a pseudogene on chromosome 1. Multiple alternatively spliced transcript variants encoding different isoforms have been identified.Specifications
| HDHD1 | |
| Monoclonal | |
| 1.2 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q08623, Q9D5U5 | |
| Pudp | |
| Full length human recombit protein of human HDHD1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI7A2 | |
| Unconjugated | |
| PUDP | |
| DXF68S1E, FAM16AX, GS1, HDHD1, HDHD1A | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 291585, 610678, 67365, 8226 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.