Learn More
HDGFRP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12380-1-AP-150UL
 View more versions of this product
Product Code. 16820074
  • 410.00 € / 150µL
Please to purchase this item. Need a web account? Register with us today!
Description
Description
HDGFRP3 enhances DNA synthesis and may play a role in cell proliferation.
Specifications
Specifications
HDGFRP3
Polyclonal
Unconjugated
HDGFL3
CGI 142, HDGF 2, HDGF2, HDGFRP3, HRP 3, HRP-3
Rabbit
Antigen Affinity Chromatography
RUO
252941, 29877, 50810
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.24 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q923W4, Q9JMG7, Q9Y3E1
HDGFL3
HDGFRP3 Fusion Protein Ag3046
150 μL
Primary
Human, Mouse, Rat