Learn More
Invitrogen™ HDGFRP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596696
Description
Immunogen sequence: EELGTLQVTS QILQKNTDVV ATLKKIRRYK ANKDVMEKAA EVYTRLKSRV LGPKIEAVQK VNKAGMEKEK AEEKLAGEEL AGEEAPQEKA EDKPSTDLSA PVNGEATSQK GESAEDKEHE EGRDSEEGPR CGSSEDLHDV REGPDLDRPG SDRQERERAR GDSEALDEES; Positive Samples: Mouse testis; Cellular Location: Nucleus.
Hepatoma-derived growth factor-related protein 2 (HDGFRP2) is a widely expressed member of protein family that contains the HDGF related proteins 1-4 and lens epithelial derived growth factor (LEDGF). These family members show extensive homology at the amino terminus (termed the hath region). Little is known of the role of HDGFRP2, but like the related LEDGF, HDGFRP2 can bind HIV-1 integrase and stimulate HIV-1 integrase activity in vitro.
Specifications
| HDGFRP2 | |
| Polyclonal | |
| Unconjugated | |
| HDGFL2 | |
| HDGF2; HDGF-2; Hdgf3; HDGF-3; Hdgfl2; HDGF-related protein 2; HDGFRP2; Hepatoma-derived growth factor 2; Hepatoma-derived growth factor 3; hepatoma-derived growth factor, related protein 2; hepatoma-derived growth factor-related protein 2; HRP2; HRP-2; UNQ785/PRO1604 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 15193, 84717 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.48 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3UMU9, Q7Z4V5 | |
| HDGFL2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 501-670 of human HDGFRP2 (NP_1160201). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.