missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDGFRP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560650
This item is not returnable.
View return policy
Description
Immunogen sequence: KVDSPDVKRC LNALEELGTL QVTSQILQKN TDVVATLKKI RRYKANKDVM EKAAEVYTRL KSRVLGPKIE AVQKV Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.
Hepatoma-derived growth factor-related protein 2 (HDGFRP2) is a widely expressed member of protein family that contains the HDGF related proteins 1-4 and lens epithelial derived growth factor (LEDGF). These family members show extensive homology at the amino terminus (termed the hath region). Little is known of the role of HDGFRP2, but like the related LEDGF, HDGFRP2 can bind HIV-1 integrase and stimulate HIV-1 integrase activity in vitro.
Specifications
| HDGFRP2 | |
| Polyclonal | |
| Unconjugated | |
| HDGFL2 | |
| HDGF2; HDGF-2; Hdgf3; HDGF-3; Hdgfl2; HDGF-related protein 2; HDGFRP2; Hepatoma-derived growth factor 2; Hepatoma-derived growth factor 3; hepatoma-derived growth factor, related protein 2; hepatoma-derived growth factor-related protein 2; HRP2; HRP-2; UNQ785/PRO1604 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84717 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z4V5 | |
| HDGFL2 | |
| Recombinant protein corresponding to Human HDGFRP2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction