missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDC Polyclonal Antibody

Rabbit Polyclonal Antibody
180.00 € - 476.00 €
Specifications
| Antigen | HDC |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17750604
![]() |
Invitrogen™
PA5119194 |
200 μL |
476.00 €
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17763314
|
Invitrogen™
PA5121918 |
20 μL |
180.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HDC Polyclonal Antibody for Western Blot, IHC (P)
The biogenic amine histamine is an important modulator of numerous physiologic processes, including neurotransmission, gastric acid secretion, and smooth muscle tone. The biosynthesis of histamine from histidine is catalyzed by the enzyme L-histidine decarboxylase (HDC; EC 4.1.1.22). This homodimeric enzyme is a pyridoxal phosphate (PLP)-dependent decarboxylase and is highly specific for its histidine substrate.Specifications
| HDC | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| Hdc | |
| AW108189; HDC; Hdc-a; Hdc-c; Hdc-e; Hdc-s; histidine decarboxylase; L-histidine decarboxylase; MGC163399 | |
| Hdc | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P19113 | |
| 3067 | |
| Recombinant protein Histidine Decarboxylase. The antigen corresponds to amino acid range 160-369 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title