Learn More
HDAC-IN-3, MedChemExpress
Product Code. p-200098708
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Product Code. 16491859
Brand: Medchem Express HY19772
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress HDAC-IN-3 is a histone deacetylase (HDAC) inhibitor, extracted from patent WO/2008040934 A1.
HDAC-IN-3 is a histone deacetylase (HDAC) inhibitor, extracted from patent WO/2008040934 A1.
Specifications
Chemical Name or Material HDAC-IN-3
Physical Form Solid
CAS 1018673-42-1
Color Earth Yellow
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C22H33N3O4
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 50 mg/mL (123.91 mM; Need ultrasonic)
SMILES O=C([C@@H](NCC1=CN=C(CCC(NO)=O)C=C1)C2CCCCC2)OC3CCCC3
Molecular Weight (g/mol) 403.52
Formula Weight 403.52
Percent Purity 99.41%
Grade Research