missing translation for 'onlineSavingsMsg'
Learn More

HC-067047, MedChemExpress

Product Code. 16492536
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16492536

Brand: Medchem Express HY100208

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress HC-067047 is a potent and selective TRPV4 antagonist and reversibly inhibits currents through the human, rat, and mouse TRPV4 orthologs with IC50 values of 48 nM, 133 nM, and 17 nM, respectively.

HC-067047 is a potent and selective TRPV4 antagonist and reversibly inhibits currents through the human, rat, and mouse TRPV4 orthologs with IC50 values of 48 nM, 133 nM, and 17 nM, respectively.

Specifications

Chemical Name or Material HC-067047
CAS 883031-03-6
Color Light Nude
Purity Grade Notes Research
Molecular Formula C26H28F3N3O2
Quantity 25 mg
Solubility Information DMSO : 50 mg/mL (106.04 mM; Need ultrasonic)
SMILES O=C(C1=C(C)N(CCCN2CCOCC2)C(C3=CC=CC=C3)=C1)NC4=CC=CC(C(F)(F)F)=C4
Molecular Weight (g/mol) 471.51
Formula Weight 471.51
Percent Purity 99.36%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.