Learn More
Lonza™ HBM™ Hepatocyte Basal Medium 500 mL
Product Code. 11675450
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11675450 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11675450 Supplier Lonza™ Supplier No. CC3197
Please to purchase this item. Need a web account? Register with us today!
Unsupplemented basal medium designed to support the growth of human hepatocytes.
  • Basal medium.
  • Contains no growth factors cytokines, or supplements.
  • Developed to support the growth of human hepatocytes.
TRUSTED_SUSTAINABILITY
Specifications
Cell Type Hepatocyte Cells
Product Type Basal Medium
Quantity 500 mL