Learn More
3M™ Harvest RC, WP6
Description
- Simplification of high cell density cell culture fluid clarification unit operations.
- Remove cells, cell debris and DNA
- Optimized for PCV high density CHO cell culture (5-8% PCV)
- Replacement of primary, secondary and guard membrane clarification stages.
- Typical product recovery of 95+% (capsules)
- Synthetic chromatographic harvest media with chemically defined extractables.
- Predictable scaling from discovery to manufacturing.
- Lower total cost of manufacturing compared to centrifugation and depth filtration.
- Lower consumption of buffer and water compared to depth filtration.
- Capsules fit into laboratory to manufacturing scale workflows.
Specifications
Specifications
| Disposable | Single-use |
| Filter Type | Filter Capsule |
| Material (Membrane) | PES |
| Sterility | Non-sterile |
| Quantity | 4/Cs. |
| Type | Chromatography Filter |