missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HARS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
149.35 € - 422.30 €
Specifications
| Antigen | HARS2 |
|---|---|
| Concentration | 0.2 mg/mL |
| Applications | Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16828283
|
Proteintech
11301-1-AP-20UL |
20 μL |
149.35 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16818283
|
Proteintech
11301-1-AP-150UL |
150 μL |
422.30 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody can bind both HARSs for the close sequences.
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. The protein encoded by this gene is an enzyme belonging to the class II family of aminoacyl-tRNA synthetases. Functioning in the synthesis of histidyl-transfer RNA, the enzyme plays an accessory role in the regulation of protein biosynthesis. The gene is located in a head-to-head orientation with HARS on chromosome five, where the homologous genes likely share a bidirectional promoter. Mutations in this gene are associated with the pathogenesis of Perrault syndrome, which involves ovarian dysgenesis and sensorineural hearing loss. Alternative splicing results in multiple transcript variants of this gene.Specifications
| HARS2 | |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Rat, Mouse | |
| P49590, Q99KK9 | |
| 23438, 307491, 70791 | |
| HARS2 Fusion Protein Ag1827 | |
| Primary | |
| -20°C | |
| HARS2 |
| 0.2 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| HARS2, HARSL, HARSR, HisRS, Histidine tRNA ligase, Histidine tRNA ligase like, HO3 | |
| Hars2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title