Learn More
HARS2 Monoclonal Antibody (OTI2D6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503794S
Description
HARS2 Monoclonal antibody specifically detects HARS2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| HARS2 | |
| Monoclonal | |
| 0.36 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P06858 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4023 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2D6 | |
| Unconjugated | |
| LPL | |
| 4631412B19Rik; D-tyrosyl-tRNA deacylase 1 homolog; EC 6.1.1.21; EGK_16929; HARS2; Hars2l; HARSL; HARSR; HARS-related; HisRS; histidine translase; histidine tRNA ligase 2, mitochondrial (putative); histidine-tRNA ligase homolog; Histidine--tRNA ligase-like; Histidyl-tRNA synthetase; histidyl-tRNA synthetase 2; histidyl-tRNA synthetase 2, mitochondrial; histidyl-tRNA synthetase 2, mitochondrial (putative); histidyl-tRNA synthetase 2, mitochondrial, hisRS; histidyl-tRNA synthetase-like; HO3; PRLTS2; Probable histidine--tRNA ligase, mitochondrial; probable histidyl-tRNA synthetase, mitochondrial; probable histidyl-tRNA synthetase, mitochondrial-like; putative histidyl-tRNA synthetase, mitochondrial; RGD1308426; zinc finger, matrin type 2; Zmat2 | |
| Human recombinant protein fragment corresponding to amino acids 28-475 of human LPL produced in HEK293T. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Das Fisher Scientific Encompass-Programm bietet Artikel an, die nicht Teil unseres Vertriebsportfolios sind. Diese Produkte verfügen in der Regel nicht über Bilder oder detaillierte Beschreibungen. Wir sind jedoch bestrebt, Ihr Einkaufserlebnis zu verbessern. Bitte verwenden Sie das untenstehende Formular, um uns Feedback zum Inhalt dieses Produkts zu geben.