Learn More
Hanna Instruments™ pH Electrode Cleaning Solution
Product Code. 10688301
Change view
Click to view available options
Capacity (Metric):
500 mL
Unit Size:
460mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10688301 500 mL 460mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10688301 Supplier Hanna Instruments™ Supplier No. HI7061L
  • ONLINE EXCLUSIVE PRICE
    33.39 €
    19.76 € / 460mL
    Save 13.63 €
    40% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Eliminates impurities and residues that are left on electrode surfaces when immersed in samples during measurement or when stored incorrectly. Hanna Instruments™ pH Electrode Cleaning Solution dissolves any mineral deposits or other coatings by soaking up to 20 minutes.
General purpose cleaning solution that can be used for a variety of applications.
  • Eliminates impurities and residues that are left on electrode surfaces when immersed in samples during measurement or when stored incorrectly.
  • Dissolves any mineral deposits or other coatings by soaking up to 20 minutes.
Specifications
Capacity (Metric) 500 mL
Filling Solution Type Aqueous
For Use With (Application) Removing impurities and residues from ph Electrodes