Learn More
Hamilton™ Needle Cleaning Concentrate
Product Code. p-183615
Click to view available options
Volume (Metric):
70 mL
500 mL
Unit Size:
Each
Product Code. 10323173
Brand: Hamilton™ 18310
Please to purchase this item. Need a web account? Register with us today!
A biodegradable, non-phosphate cleaning concentrate that is suitable for many common sample types.
  • Non-alkaline, non-phosphate and non-detergent based
  • 70mL or 500mL bottle
  • For syringes and needles
Specifications
Product Type Cleaning Concentrate
For Use With (Equipment) Syringe and Needle Cleaning Kit
Volume (Metric) 70 mL