Learn More
Hamilton™ Inert Luer Syringe Gas Sampling Valve
Description
Kel-f hub needles are able to attach to the valve. This valve is pressure tested a 100 psi.
Specifications
Specifications
| Bore Size (English) | 0.039 in. |
| For Use With (Application) | Storing and transporting samples in any Gastight™ TLL syringe |
| Type | Inert Gas Sampling Valve |