Learn More
O&M Halyard Quick Check Sterilization Wrap (H500)
GREENER_CHOICE
Product Code. 30274732 Shop All Halyard Products
Click to view available options
Dimensions (L x W):
114 x 114 cm (45 x 45 in.)
121 x 121 cm (48 x 48 in.)
137 x 137 cm (54 x 54 in.)
137 x 182 cm (54 x 72 in.)
45 x 45 cm (18 x 18 in.)
60 x 60 cm (24 x 24 in.)
76 x 76 cm (30 x 30 in.)
91 x 91 cm (36 x 36 in.)
Unit Size:
Pack of 120
Pack of 24
Pack of 288
Pack of 48
Pack of 72
Product Code. 30274732
Brand: O&M 34199
Only null left
Add to basket
Add to basket
Using Halyard Quick Check wraps is a fast, easy way to ensure instrument sterility is uncompromised.
  • Inspect trays easier and faster than ever
  • Superior microbial barrier protection with patented Powerguard technology
  • Cleared for all of the same sterilization systems as Halyard Kimguard One-step Wrap
  • Made from recyclable No. 5 polypropylene fabric
  • Made in the USA
Specifications
Autoclavable Autoclavable
Certifications/Compliance Meets NFPA Class 1
Color Blue
For Use With (Equipment) Heavy instrument sets, retractors, and applications where frequent handling occurs (13 to 17 lb.)
Material SMS Fabric (H500)
Sterility Non-sterile
Latex Free Yes
Dimensions (L x W) 91 x 91 cm (36 x 36 in.)
Length (English) 36 in.
Length (Metric) 91 cm
Width (English) 36 in.
Width (Metric) 91 cm
Product Type Sterilization Wrap