Learn More
MedChemExpress HA Peptide
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Purity | 99.23% |
| CAS | 92000-76-5 |
| Content And Storage | Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month |
| For Use With (Application) | COVID-19-immunoregulation |
| Molecular Weight (g/mol) | 1102.15 g/mol |
| Product Type | Natural Products |
| Quantity | 10 mg |
| Shipping Condition | Room temperature |
| Solubility | H2O : 50 mg/mL (45.37 mM∣Need ultrasonic) |