Learn More
Mettler Toledo™ HA 405-DXK-S8 pH Combination Electrode
Product Code. 10140641 Shop All Mettler Toledo Products
Change view
Click to view available options
Length (Metric):
120 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
10140641 120 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10140641 Supplier Mettler Toledo™ Supplier No. 59903146
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Used for a variety of tough chemical applications. Mettler Toledo™ HA 405-DXK-S8 pH Combination Electrode is a pressure-resistant, low-maintenance pH-electrode
  • Low maintenance - A solid polymer electrolyte (no refilling and pressurizing of electrolyte) in direct contact with the process media via an open junction. No clogging in contaminated media due to the open junction.
  • Long operational - electrode lifetime The electrode allows reliable process control in highly demanding chemical processes, particulary suitable for protein-containing solutions, emulsions and suspensions, hot caustic solutions, sulfide-bearing media etc..
  • High pressure resistance - The electrode covers a generous pressure range and shows high resistance to process pressure fluctuations
TRUSTED_SUSTAINABILITY
Specifications
Type pH Combination Electrode
Length (Metric) 120 mm