missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ H3K27M Recombinant Rabbit Monoclonal Antibody (35C11)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550100
This item is not returnable.
View return policy
Description
H3K27M Recombinant Monoclonal Antibody for IHC (P), ELISA
Core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling.
Specifications
| H3K27M | |
| Recombinant Monoclonal | |
| 1.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P68431 | |
| HIST1H3A | |
| A synthesized peptide derived from Human HIST1H3A. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| 35C11 | |
| Unconjugated | |
| HIST1H3A | |
| H3FA; H3FB; H3FC; H3FD; H3FF; H3FH; H3FI; H3FJ; H3FK; H3FL; HIST1H3A; HIST1H3B; HIST1H3C; HIST1H3D; HIST1H3E; HIST1H3F; HIST1H3G; HIST1H3H; HIST1H3I; HIST1H3J | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 8350 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction