Learn More
H-L-MeThr-OH, TRC
Product Code. 30481415
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481415 100 mg 100mg
30474549 10 mg 10mg
30476251 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30481415 Supplier Toronto Research Chemicals Supplier No. TRCM222748100MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2812-28-4
Molecular Formula C5H11NO3
Molecular Weight (g/mol) 133.15
Synonym N-Methyl-L-threonine, N-Methylthreonine
IUPAC Name (2S,3R)-3-hydroxy-2-(methylamino)butanoic acid
SMILES CN[C@@H]([C@@H](C)O)C(=O)O
Specifications
InChI Formula InChI=1S/C5H11NO3/c1-3(7)4(6-2)5(8)9/h3-4,6-7H,1-2H3,(H,8,9)/t3-,4+/m1/s1
Quantity 100 mg
Formula Weight 133.0739
Shipping Condition Room Temperature
Chemical Name or Material H-L-MeThr-OH