Learn More
H-Glu(OEt)-OH, TRC
Product Code. 30393608
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30393608 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30393608 Supplier Toronto Research Chemicals Supplier No. TRCG598193500MG
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material H-Glu(OEt)-OH
CAS 1119-33-1
Molecular Formula C7H13NO4
Quantity 500 mg
Synonym 5-Ethyl L-Glutamate, Ethyl L-Glutamate, L-Glutamic Acid gamma-Ethyl Ester, NSC 156977, NSC 16885, gamma-Ethyl L-Glutamate, gamma-Ethyl Glutamate, gamma-Monoethyl Glutamate
InChI Formula InChI=1S/C7H13NO4/c1-2-12-6(9)4-3-5(8)7(10)11/h5H,2-4,8H2,1H3,(H,10,11)/t5-/m0/s1
SMILES CCOC(=O)CC[C@H](N)C(=O)O
IUPAC Name (2S)-2-amino-5-ethoxy-5-oxopentanoic acid
Molecular Weight (g/mol) 175.18
Formula Weight 175.0845