Learn More
H-Glu(OEt)-OH, TRC
Product Code. 30403204
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30403204 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30403204 Supplier Toronto Research Chemicals Supplier No. TRCG5981932.5G
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material H-Glu(OEt)-OH
CAS 1119-33-1
Molecular Formula C7H13NO4
Quantity 2.5 g
Synonym 5-Ethyl L-Glutamate, Ethyl L-Glutamate, L-Glutamic Acid gamma-Ethyl Ester, NSC 156977, NSC 16885, gamma-Ethyl L-Glutamate, gamma-Ethyl Glutamate, gamma-Monoethyl Glutamate
InChI Formula InChI=1S/C7H13NO4/c1-2-12-6(9)4-3-5(8)7(10)11/h5H,2-4,8H2,1H3,(H,10,11)/t5-/m0/s1
SMILES CCOC(=O)CC[C@H](N)C(=O)O
IUPAC Name (2S)-2-amino-5-ethoxy-5-oxopentanoic acid
Molecular Weight (g/mol) 175.18
Formula Weight 175.0845